Merck
  • Oral Nanostructured Lipid Carriers Loaded with Near-Infrared Dye for Image-Guided Photothermal Therapy.
Oral Nanostructured Lipid Carriers Loaded with Near-Infrared Dye for Image-Guided Photothermal Therapy.
ACS applied materials & interfaces (2016-09-15)
Gang Chen, Kaikai Wang, Yiwen Zhou, Ling Ding, Aftab Ullah, Qi Hu, Minjie Sun, David Oupický
ABSTRACT
Photothermal therapy exerts its anticancer effect by converting laser radiation energy into hyperthermia using a suitable photosensitizer. This study reports development of nanostructured lipid carriers (NLCs) suitable for noninvasive oral delivery of a near-infrared photosensitizer dye IR780. The carrier encapsulating the dye (IR780@NLCs) was stable in simulated gastric and intestinal conditions and showed greatly enhanced oral absorption of IR780 when compared with the free dye. As a result of increased oral bioavailability, enhanced accumulation of the dye in subcutaneous mouse colon tumors (CT-26 cells) was observed following oral gavage of IR780@NLCs. Photothermal antitumor activity of orally administered IR780@NLCs was evaluated following local laser irradiation of the CT-26 tumors. We observed significant effect of the photothermal IR780@NLCs treatment on the rate of the tumor growth and no toxicity associated with the oral administration of IR780@NLCs. Overall, orally administered IR780@NLCs represents a safe and noninvasive method to achieve systemic tumor delivery of a photosensitizing dye for applications in photothermal anticancer therapies.
Product Description
Trilaurin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.