Merck
  • Targeting methyltransferase PRMT5 eliminates leukemia stem cells in chronic myelogenous leukemia.
Targeting methyltransferase PRMT5 eliminates leukemia stem cells in chronic myelogenous leukemia.
The Journal of clinical investigation (2016-09-20)
Yanli Jin, Jingfeng Zhou, Fang Xu, Bei Jin, Lijing Cui, Yun Wang, Xin Du, Juan Li, Peng Li, Ruibao Ren, Jingxuan Pan
ABSTRACT
Imatinib-insensitive leukemia stem cells (LSCs) are believed to be responsible for resistance to BCR-ABL tyrosine kinase inhibitors and relapse of chronic myelogenous leukemia (CML). Identifying therapeutic targets to eradicate CML LSCs may be a strategy to cure CML. In the present study, we discovered a positive feedback loop between BCR-ABL and protein arginine methyltransferase 5 (PRMT5) in CML cells. Overexpression of PRMT5 was observed in human CML LSCs. Silencing PRMT5 with shRNA or blocking PRMT5 methyltransferase activity with the small-molecule inhibitor PJ-68 reduced survival, serial replating capacity, and long-term culture-initiating cells (LTC-ICs) in LSCs from CML patients. Further, PRMT5 knockdown or PJ-68 treatment dramatically prolonged survival in a murine model of retroviral BCR-ABL-driven CML and impaired the in vivo self-renewal capacity of transplanted CML LSCs. PJ-68 also inhibited long-term engraftment of human CML CD34+ cells in immunodeficient mice. Moreover, inhibition of PRMT5 abrogated the Wnt/β-catenin pathway in CML CD34+ cells by depleting dishevelled homolog 3 (DVL3). This study suggests that epigenetic methylation modification on histone protein arginine residues is a regulatory mechanism to control self-renewal of LSCs and indicates that PRMT5 may represent a potential therapeutic target against LSCs.
Sigma-Aldrich
Anti-PRMT5 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, clone 62-141-13, Upstate®, from rabbit
Sigma-Aldrich
Anti-Kip2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-dimethyl Histone H3 (Arg2), symmetric Antibody, serum, from rabbit
Sigma-Aldrich
Anti-STAT3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-phospho-STAT3 (Tyr705) Antibody, clone 9E12, clone 9E12, Upstate®, from mouse
Sigma-Aldrich
Anti-WDR5 Antibody, Upstate®, from rabbit