Merck
  • In vitro antiproliferative and antioxidant effects of urolithin A, the colonic metabolite of ellagic acid, on hepatocellular carcinomas HepG2 cells.
In vitro antiproliferative and antioxidant effects of urolithin A, the colonic metabolite of ellagic acid, on hepatocellular carcinomas HepG2 cells.
Toxicology in vitro : an international journal published in association with BIBRA (2015-04-26)
Yun Wang, Zhenpeng Qiu, Benhong Zhou, Cong Liu, Jinlan Ruan, Qiujin Yan, Jianming Liao, Fan Zhu
ABSTRACT
The intestinal metabolites of ellagic acid (EA), urolithins are known to effectively inhibit cancer cell proliferation. This study investigates antiproliferative and antioxidant effects of urolithin A (UA) on cell survival of the HepG2 hepatic carcinomas cell line. The antiproliferative effects of UA (0-500 μM) on HepG2 cells were determined using a CCK assay following 12-36 h exposure. Effects on β-catenin and other factors of expression were assessed by using real-time PCR and Western blot. We found that UA showed potent antiproliferative activity on HepG2 cells. When cell death was induced by UA, it was found that the expression of β-catenin, c-Myc and Cyclin D1 were decreased and TCF/LEF transcriptional activation was notably down-regulated. UA also increased protein expression of p53, p38-MAPK and caspase-3, but suppressed expression of NF-κB p65 and other inflammatory mediators. Furthermore, the antioxidant assay afforded by UA and EA treatments was associated with decreases in intracellular ROS levels, and increases in intracellular SOD and GSH-Px activity. These results suggested that UA could inhibit cell proliferation and reduce oxidative stress status in liver cancer, thus acting as a viably effective constituent for HCC prevention and treatment.
Sigma-Aldrich
Urolithin A, ≥97% (HPLC)