Merck
  • A caffeine-sensitive membrane electrode: previous misleading report and present approach.
A caffeine-sensitive membrane electrode: previous misleading report and present approach.
Analytica chimica acta (2008-06-19)
Takashi Katsu, Yumi Tsunamoto, Nobumitsu Hanioka, Keiko Komagoe, Kazufumi Masuda, Shizuo Narimatsu
ABSTRACT
Although a previous study [S.S.M. Hassan, M.A. Ahmed, M.M. Saoudi, Anal. Chem. 57 (1985) 1126] had shown that a caffeine-sensitive electrode made with picrylsulfonate and 1-octanol as a cation-exchanger and a solvent mediator, respectively, had a wide working pH range (5.5-9.5) and exhibited a Nernstian response, we could not find such response in this electrode. The present result was reasonable, because the pK(a) value of caffeinium ion was reported to be around 0.7 and the neutral form of caffeine was predominant in the pH range examined. Thus, we reinvestigated the response characteristics of a caffeine electrode, taking into consideration the pK(a) value, and constructed a new electrode with a combination of the lipophilic cation-exchanger, tetrakis[3,5-bis(2-methoxyhexafluoro-2-propyl)phenyl]borate (HFPB), and the solvent mediator with high degree of dielectric constant, 2-fluoro-2'-nitrodiphenyl ether (FNDPE). This electrode showed a pH-dependent response to caffeinium ion and gave a detection limit of 50muM with a slope of 55mV per concentration decade at pH 2. The use of other solvent mediators was less effective than that of FNDPE. The electrode was applied for the determination of caffeine in some central stimulants.
Product Description
Supelco
Sodium tetrakis[3,5-bis(1,1,1,3,3,3-hexafluoro-2-methoxy-2-propyl)phenyl]borate trihydrate, Selectophore, for ion-selective electrodes
Pricing and availability is not currently available.