Merck
  • In-situ photocrosslinkable nanohybrid elastomer based on polybutadiene/polyhedral oligomeric silsesquioxane.
In-situ photocrosslinkable nanohybrid elastomer based on polybutadiene/polyhedral oligomeric silsesquioxane.
Materials science & engineering. C, Materials for biological applications (2016-08-16)
Seyed Amin Mirmohammadi, Mehdi Nekoomanesh-Haghighi, Somayyeh Mohammadian Gezaz, Naeimeh Bahri-Laleh, Mohammad Atai
ABSTRACT
Hydroxyl functionalized nano-sized POSS or ethyleneglycol as diol monomers was incorporated to hydroxyl-terminated polybutadiene (HTPBD) chain in the presence of fumaryl chloride as extender. Blue light photocrosslinking system based on camphorquinone (photoinitiator) and dimethylaminoethyl methacrylate (accelerator) was applied to cure these two synthesized fumarate based macromers. Self-crosslinkability of unsaturated macromers and also crosslinking in presence of a reactive diluent were investigated in absence and presence of 1,4-butanediol dimethacrylate, respectively. Finally, photocured samples were characterized by XRD, SEM, equilibrium swelling study, TGA, DMTA, AFM and cell culture. The results showed that incorporation of POSS nanoparticle into the polymer matrix with a perfect distribution and dispersion can enhance thermal stability, mechanical and biocompatibility properties which can prove a good potential of this in-situ photocrosslinkable nanohybrid in medical applications.
Sigma-Aldrich
Fumaryl chloride, 95%
Pricing and availability is not currently available.