Merck
  • Role of the JP45-Calsequestrin Complex on Calcium Entry in Slow Twitch Skeletal Muscles.
Role of the JP45-Calsequestrin Complex on Calcium Entry in Slow Twitch Skeletal Muscles.
The Journal of biological chemistry (2016-05-18)
Barbara Mosca, Jan Eckhardt, Leda Bergamelli, Susan Treves, Rossana Bongianino, Marco De Negri, Silvia G Priori, Feliciano Protasi, Francesco Zorzato
ABSTRACT
We exploited a variety of mouse models to assess the roles of JP45-CASQ1 (CASQ, calsequestrin) and JP45-CASQ2 on calcium entry in slow twitch muscles. In flexor digitorum brevis (FDB) fibers isolated from JP45-CASQ1-CASQ2 triple KO mice, calcium transients induced by tetanic stimulation rely on calcium entry via La(3+)- and nifedipine-sensitive calcium channels. The comparison of excitation-coupled calcium entry (ECCE) between FDB fibers from WT, JP45KO, CASQ1KO, CASQ2KO, JP45-CASQ1 double KO, JP45-CASQ2 double KO, and JP45-CASQ1-CASQ2 triple KO shows that ECCE enhancement requires ablation of both CASQs and JP45. Calcium entry activated by ablation of both JP45-CASQ1 and JP45-CASQ2 complexes supports tetanic force development in slow twitch soleus muscles. In addition, we show that CASQs interact with JP45 at Ca(2+) concentrations similar to those present in the lumen of the sarcoplasmic reticulum at rest, whereas Ca(2+) concentrations similar to those present in the SR lumen after depolarization-induced calcium release cause the dissociation of JP45 from CASQs. Our results show that the complex JP45-CASQs is a negative regulator of ECCE and that tetanic force development in slow twitch muscles is supported by the dynamic interaction between JP45 and CASQs.
Sigma-Aldrich
Anti-Calsequestrin-1 (N-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Protein G−Peroxidase from Streptococcus sp., recombinant, expressed in unspecified host, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution