Merck
Analysis of focal adhesions and cytoskeleton by custom microarray.
Methods in molecular biology (Clifton, N.J.) (2007-04-10)
Matthew J Dalby, Stephen J Yarwood
ABSTRACT
Focal adhesions and the cell cytoskeleton (intermediate filaments, microfilaments, microtubules) are involved in mechanotransduction-both direct (transduction of mechanical forces to the nucleus) and indirect (transduction of chemical signaling cascades to the nucleus). Thus, observation of changes in focal adhesion and cytoskeletal organization can be invaluable in research such as drug treatments and medical material testing in vitro. Here we describe how to stain human fibroblasts for vinculin (located to focal adhesions), actin (microfilaments), tubulin (microtubules), and vimentin (intermediate filaments) and how to perform custom microarray experiments. Comparative analysis of the immunofluorescence and array data should allow the researcher to build up a global picture of changes to both direct and indirect mechanotransduction through the cytoskeleton from focal adhesions.
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, DNA, Cytoskeleton Genes
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock, Cytoskeleton Genes
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Cytoskeleton Genes