- Synthesis and structure-activity study of protease inhibitors. V. Chemical modification of 6-amidino-2-naphthyl 4-guanidinobenzoate.
Synthesis and structure-activity study of protease inhibitors. V. Chemical modification of 6-amidino-2-naphthyl 4-guanidinobenzoate.
By developing 6-amidino-2-naphthyl 4-guanidinobenzoate (I, FUT-175) as a basic structure, its various derivatives were synthesized and their inhibitory activities on trypsin, plasmin, kallikrein, thrombin, C1r and C1s as well as on complement-mediated hemolysis were examined. The protective effect of these compounds on complement-mediated Forssman shock was also examined in guinea pigs. 6-Amidino-2-naphthyl 4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-benzoate (41, FUT-187) was found to be a suitable compound for oral administration with anti-complement activity superior to that of compound I.