Merck
  • Des-aspartate-angiotensin-I and angiotensin IV improve glucose tolerance and insulin signalling in diet-induced hyperglycaemic mice.
Des-aspartate-angiotensin-I and angiotensin IV improve glucose tolerance and insulin signalling in diet-induced hyperglycaemic mice.
Biochemical pharmacology (2011-08-02)
Yong-Chiat Wong, Meng-Kwoon Sim, Kok-Onn Lee
ABSTRACT
Although clinical studies suggested that blockade of the renin-angiotensin system may prevent diabetes, the mechanism is uncertain. As a follow-up to an earlier study, we investigated how des-aspartate-angiotensin-1 (DAA-1) and its metabolite, angiotensin IV (Ang-IV) improved glucose tolerance in diet-induced hyperglycaemic mice. Male C57BL/6J mice were fed a high-fat-high-sucrose (HFD) or normal (ND) diet for 52 weeks. HFD animals were orally administered either DAA-I (600nmol/kg/day), Ang-IV (400nmol/kg/day) or distilled water. Body weight, blood glucose and insulin were measured fortnightly. Inflammatory and insulin signalling transducers that are implicated in hyperglycaemia were analyzed in skeletal muscles at 52 weeks. HFD animals developed hyperglycemia, hyperinsulinemia and obesity. DAA-I and Ang-IV improved glucose tolerance but had no effect on hyperinsulinemia and obesity. Skeletal muscles of HFD animals showed increased level of ROS, gp91 of NADPH oxidase, pJNK and AT(1)R-JAK-2-IRS-1 complex. Both DAA-I and Ang-IV attenuated these increases. Insulin-induced activation of IR, IRS-1, IRS-1-PI3K coupling, phosphorylation of Akt, and GLUT4 translocation were attenuated in skeletal muscles of HFD animals. The attenuation was significantly ameliorated in DAA-I-treated HFD animals. In corresponding Ang-IV treated animals, insulin induced IRAP and PI3K interaction, activation of pAkt and GLUT4 translocation, but no corresponding activation of IR, IRS-1 and IRS-1-PI3K coupling were observed. DAA-I and Ang-IV improved glucose tolerance, insulin signalling, and para-inflammatory processes linked to hyperglycaemia. DAA-I acts via the angiotensin AT(1) receptor and activates the insulin pathway. Ang-IV acts via IRAP, which couples PI3K and activates the later part of the insulin pathway.