Merck
  • Ph3As-catalyzed wittig-type olefination of aldehydes with diazoacetate in the presence of Na2S2O4.
Ph3As-catalyzed wittig-type olefination of aldehydes with diazoacetate in the presence of Na2S2O4.
The Journal of organic chemistry (2007-07-28)
Peng Cao, Chuan-Ying Li, Yan-Biao Kang, Zuowei Xie, Xiu-Li Sun, Yong Tang
ABSTRACT
In the presence of sodium hydrosulfite and a catalytic amount of AsPh3 and Fe(TCP)Cl, aldehydes react with ethyl diazoacetate to give the corresponding alpha,beta-unsaturated esters in high yields with excellent stereoselectivities (E/Z > 50/1).
Sigma-Aldrich
4-(Trifluoromethyl)benzaldehyde, 98%
Pricing and availability is not currently available.