Merck
A concise asymmetric synthesis of torcetrapib.
The Journal of organic chemistry (2007-07-13)
Meritxell Guinó, Pim Huat Phua, Jean-Claude Caille, King Kuok Mimi Hii
ABSTRACT
Optically active torcetrapib was synthesized in seven steps from achiral precursors without the need for protecting groups, utilizing an enantioselective aza-Michael reaction to achieve asymmetry.
Sigma-Aldrich
4-(Trifluoromethyl)aniline, 99%