Merck
  • Microemulsion electrokinetic chromatography in suppressed electroosmotic flow environment. Separation of fat-soluble vitamins.
Microemulsion electrokinetic chromatography in suppressed electroosmotic flow environment. Separation of fat-soluble vitamins.
Journal of chromatography. A (2000-05-24)
S Pedersen-Bjergaard, O Naess, S Moestue, K E Rasmussen
ABSTRACT
Microemulsion electrokinetic chromatography (MEEKC) was carried out in a pH 2.5 phosphate buffer to effectively suppress the electroosmotic flow (EOF). With 66.6% (w/w) 25 mM phosphate buffer pH 2.5, 20.0% (w/w) 2-propanol, 6.6% (w/w) 1-butanol, 6.0% (w/w) sodium lauryl sulphate (SDS), and 0.8% (w/w) n-octane as the separation medium, the fat-soluble vitamins A palmitate, E acetate, and D3 were baseline separated within 11 min. With strongly suppressed EOF, the polarity of the separation voltage was reversed (positive electrode at the outlet); the n-octane micro droplets surrounded by negatively charged SDS molecules migrated towards the detector. The aqueous part of the microemulsion was modified with 20% (w/w) 2-propanol to improve partition between the n-octane phase and the surrounding aqueous medium. The fat-soluble vitamins were separated in order of decreasing hydrophobicity with a high migration time stability (repeatable within 0.1% RSD). Excellent accuracy and precision were obtained when the system was applied for the determination of vitamin E acetate in commercial vitamin tablets; quantitative data corresponded to 97.0% of label claim, intra-day results varied within 1.72% RSD (n=6), and inter-day results varied within 3.22% RSD (n=5).
Product Description
Supelco
1-Butanol, analytical standard
Pricing and availability is not currently available.
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.