Merck
  • A highly selective, organocatalytic route to chiral 1,2-oxazines from ketones.
A highly selective, organocatalytic route to chiral 1,2-oxazines from ketones.
Chemical communications (Cambridge, England) (2006-10-10)
Sirirat Kumarn, David M Shaw, Steven V Ley
ABSTRACT
A sequential, organocatalysed asymmetric reaction to access chiral 1,2-oxazines from achiral ketone starting materials is reported, which proceeds in moderate to good yields and excellent enantioselectivity.
Sigma-Aldrich
(S)-(–)-5-(2-Pyrrolidinyl)-1H-tetrazole, 96%