Merck
  • An Inducible System for Rapid Degradation of Specific Cellular Proteins Using Proteasome Adaptors.
An Inducible System for Rapid Degradation of Specific Cellular Proteins Using Proteasome Adaptors.
PloS one (2016-04-05)
Shameika R Wilmington, Andreas Matouschek
ABSTRACT
A common way to study protein function is to deplete the protein of interest from cells and observe the response. Traditional methods involve disrupting gene expression but these techniques are only effective against newly synthesized proteins and leave previously existing and stable proteins untouched. Here, we introduce a technique that induces the rapid degradation of specific proteins in mammalian cells by shuttling the proteins to the proteasome for degradation in a ubiquitin-independent manner. We present two implementations of the system in human culture cells that can be used individually to control protein concentration. Our study presents a simple, robust, and flexible technology platform for manipulating intracellular protein levels.
Sigma-Aldrich
GA3-AM, ≥95% (HPLC)
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
ANTI-FLAG® BioM2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution