Merck
  • Glycine N-acyltransferase-like 3 is responsible for long-chain N-acylglycine formation in N18TG2 cells.
Glycine N-acyltransferase-like 3 is responsible for long-chain N-acylglycine formation in N18TG2 cells.
Journal of lipid research (2016-03-27)
Kristen A Jeffries, Daniel R Dempsey, Emma K Farrell, Ryan L Anderson, Gabrielle J Garbade, Tatyana S Gurina, Imran Gruhonjic, Carly A Gunderson, David J Merkler
ABSTRACT
Long-chain fatty acid amides are signaling lipids found in mammals and other organisms; however, details of the metabolic pathways for the N-acylglycines and primary fatty acid amides (PFAMs) have remained elusive. Heavy-labeled precursor and subtraction lipidomic experiments in mouse neuroblastoma N18TG2 cells, a model cell line for the study of fatty acid amide metabolism, establish the biosynthetic pathways for the N-acylglycines and the PFAMs. We provide evidence that the N-acylglycines are formed by a long-chain specific glycine-conjugating enzyme, glycine N-acyltransferase-like 3 (GLYATL3). siRNA knockdown of GLYATL3 in the N18TG2 cells resulted in a decrease in the levels of the N-acylglycines and the PFAMs. This is the first report of an enzyme responsible for long-chain N-acylglycine production in cellula. The production of the PFAMs in N18TG2 cells was reported to occur by the oxidative cleavage of the N-acylglycines, as catalyzed by peptidylglycine α-amidating monooxygenase (PAM). siRNA knockdown of PAM resulted in an accumulation of [(13)C18]N-oleoylglycine and decreased levels of [(13)C18]oleamide when the N18TG2 cells were grown in the presence of [(13)C18]oleic acid. The addition of [1-(13)C]palmitate to the N18TG2 cell growth media led to the production of a family of [1-(13)C]palmitoylated fatty acid amides, consistent with the biosynthetic pathways detailed herein.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
PalGly, ≥98% (HPLC), solid