Merck
  • Design, synthesis, and evaluation of novel biarylpyrimidines: a new class of ligand for unusual nucleic acid structures.
Design, synthesis, and evaluation of novel biarylpyrimidines: a new class of ligand for unusual nucleic acid structures.
Journal of medicinal chemistry (2006-08-18)
Richard T Wheelhouse, Sharon A Jennings, Victoria A Phillips, Dimitrios Pletsas, Peter M Murphy, Nichola C Garbett, Jonathan B Chaires, Terence C Jenkins
ABSTRACT
Biarylpyrimidines are characterized as selective ligands for higher-order nucleic acid structures. A concise and efficient synthesis has been devised incorporating Suzuki biaryl cross-coupling of dihalopyrimidines. Two ligand series are described based on the parent thioether 4,6-bis[4-[[2-(dimethylamino)ethyl]mercapto]phenyl]pyrimidine (1a) and amide 4,6-bis(4[(2-(dimethylamino)ethyl)carboxamido]phenyl)pyrimidine (2a) compounds. In UV thermal denaturation studies with the poly(dA) x [poly(dT)]2 triplex structure, thioethers showed stabilization of the triplex form (Delta Tm < or = 20 degrees C). In contrast, amides showed duplex stabilization (Delta Tm < or = 15 degrees C) and either negligible stabilization or specific destabilization (Delta Tm = -5 degrees C) of the triplex structure. Full spectra of nucleic acid binding preferences were determined by competition dialysis. The strongest interacting thioether bound preferentially to the poly(dA) x [poly(dT)]2 triplex, K(app) = 1.6 x 10(5) M(-1) (40 x K(app) for CT DNA duplex). In contrast, the strongest binding amide selected the (T2G20T2)4 quadruplex structure, K(app) = 0.31 x 10(5) M(-1) (6.5 x K(app) for CT DNA duplex).
Sigma-Aldrich
4,6-Dichloropyrimidine, 97%
Pricing and availability is not currently available.