Merck
  • High-Level HOOK3 Expression Is an Independent Predictor of Poor Prognosis Associated with Genomic Instability in Prostate Cancer.
High-Level HOOK3 Expression Is an Independent Predictor of Poor Prognosis Associated with Genomic Instability in Prostate Cancer.
PloS one (2015-08-01)
Nathaniel Melling, Levon Harutyunyan, Claudia Hube-Magg, Martina Kluth, Ronald Simon, Patrick Lebok, Sarah Minner, Maria Christina Tsourlakis, Christina Koop, Markus Graefen, Meike Adam, Alexander Haese, Corinna Wittmer, Stefan Steurer, Jakob Izbicki, Guido Sauter, Waldemar Wilczak, Thorsten Schlomm, Till Krech
ABSTRACT
Hook microtubule-tethering protein 3 (HOOK3) is an adaptor protein for microtubule-dependent intracellular vesicle and protein trafficking. In order to assess the role of HOOK3 in prostate cancer we analyzed HOOK3 expression by immunohistochemistry on a TMA containing more than 12,400 prostate cancers. Results were compared to tumor phenotype and PSA recurrence as well as aberrations possibly defining relevant molecular subtypes such as ERG status and deletions of 3p13, 5q21, 6q15 and PTEN. HOOK3 immunostaining was negative in normal luminal cells of prostate epithelium, whereas 53.3% of 10,572 interpretable cancers showed HOOK3 expression, which was considered low in 36.4% and high in 16.9% of cases. High-level HOOK3 expression was linked to advanced tumor stage, high Gleason score, high proliferation index, positive lymph node stage, and PSA recurrence (p<0.0001 each). The prognostic role of HOOK3 expression was independent of established clinico-pathological parameters both in preoperative and postoperative settings. Comparisons with molecular features were performed to draw conclusions on the potential function of HOOK3 in the prostate. A strong association with all examined deletions is consistent with a role of HOOK3 for maintaining genomic integrity by contributing to proper centrosome assembly. Finding HOOK3 expression in 74% of ERG positive but in only 38% of ERG negative cancers (p<0.0001) further suggests functional interactions between these genes. In conclusion, the results of our study identify HOOK3 as a strong candidate prognostic marker with a possible role in maintaining genomic integrity in prostate cancer, which may have potential for inclusion into clinical routine assays.
Sigma-Aldrich
Anti-HOOK3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.