Merck
  • Development of a Method for the N-Arylation of Amino Acid Esters with Aryl Triflates.
Development of a Method for the N-Arylation of Amino Acid Esters with Aryl Triflates.
Organic letters (2016-08-09)
Sandra M King, Stephen L Buchwald
ABSTRACT
A general method for the N-arylation of amino acid esters with aryl triflates is described. Both α- and β-amino acid esters, including methyl, tert-butyl, and benzyl esters, are viable substrates. Reaction optimization was carried out by design of experiment (DOE) analysis using JMP software. The mild reaction conditions, which use t-BuBrettPhos Pd G3 or G4 precatalyst, result in minimal racemization of the amino acid ester. This method is the first synthetic application of the t-BuBrettPhos Pd G4 precatalyst. Mechanistic studies show that the observed erosion in enantiomeric excess is due to racemization of the amino acid ester starting material and not of the product.
Sigma-Aldrich
rac-BINAP Pd G4
Pricing and availability is not currently available.
Sigma-Aldrich
tBuBrettPhos Pd G3, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
XantPhos Pd G4
Pricing and availability is not currently available.