Merck
  • A genetic map of Peromyscus with chromosomal assignment of linkage groups (a Peromyscus genetic map).
A genetic map of Peromyscus with chromosomal assignment of linkage groups (a Peromyscus genetic map).
Mammalian genome : official journal of the International Mammalian Genome Society (2014-01-22)
Jane Kenney-Hunt, Adrienne Lewandowski, Travis C Glenn, Julie L Glenn, Olga V Tsyusko, Rachel J O'Neill, Judy Brown, Clifton M Ramsdell, Quang Nguyen, Tony Phan, Kimberly R Shorter, Michael J Dewey, Gabor Szalai, Paul B Vrana, Michael R Felder
ABSTRACT
The rodent genus Peromyscus is the most numerous and species-rich mammalian group in North America. The naturally occurring diversity within this genus allows opportunities to investigate the genetic basis of adaptation, monogamy, behavioral and physiological phenotypes, growth control, genomic imprinting, and disease processes. Increased genomic resources including a high quality genetic map are needed to capitalize on these opportunities. We produced interspecific hybrids between the prairie deer mouse (P. maniculatus bairdii) and the oldfield mouse (P. polionotus) and scored meiotic recombination events in backcross progeny. A genetic map was constructed by genotyping of backcross progeny at 185 gene-based and 155 microsatellite markers representing all autosomes and the X-chromosome. Comparison of the constructed genetic map with the molecular maps of Mus and Rattus and consideration of previous results from interspecific reciprocal whole chromosome painting allowed most linkage groups to be unambiguously assigned to specific Peromyscus chromosomes. Based on genomic comparisons, this Peromyscus genetic map covers ~83% of the Rattus genome and 79% of the Mus genome. This map supports previous results that the Peromyscus genome is more similar to Rattus than Mus. For example, coverage of the 20 Rattus autosomes and the X-chromosome is accomplished with only 28 segments of the Peromyscus map, but coverage of the 19 Mus autosomes and the X-chromosome requires 40 chromosomal segments of the Peromyscus map. Furthermore, a single Peromyscus linkage group corresponds to about 91% of the rat and only 76% of the mouse X-chromosomes.
Product Description
Roche
Nick Translation Mix, sufficient for 50 labeling reactions, pkg of 200 μL, solution
Pricing and availability is not currently available.