Merck
  • Highly enantioselective alpha-aminoxylation of aldehydes and ketones in ionic liquids.
Highly enantioselective alpha-aminoxylation of aldehydes and ketones in ionic liquids.
The Journal of organic chemistry (2006-10-10)
Kun Huang, Zhi-Zhen Huang, Xin-Liang Li
ABSTRACT
As the first example for the synthesis of optically active alpha-hydroxyaldehydes and alpha-hydroxyketones in ionic liquids, we applied RTILs into L-proline catalyzed direct enantioselective alpha-aminoxylation of both aldehydes and ketones successfully. This protocol features a number of advantages, such as recycling of green solvents and chiral organocatalyst, high yields, excellent enantioselectivities, short reaction times, and broad substrate scope.
Sigma-Aldrich
Tetrahydro-4H-pyran-4-one, 99%
Pricing and availability is not currently available.