- Evaluation of eye irritation by S-(-)-10,11-dihydroxyfarnesic acid methyl ester secreted by
Evaluation of eye irritation by S-(-)-10,11-dihydroxyfarnesic acid methyl ester secreted by
Experimental and therapeutic medicine (2013-10-19)
Hyeong-U Son, Sang-Han Lee
PMID24137288
ABSTRACT
The aim of this study was to investigate whether S-(-)-10,11-dihydroxyfarnesic acid methyl ester produced by cell subtype