Merck
  • Determination of nitrogen mustard hydrolysis products, ethanolamines by gas chromatography-mass spectrometry after tert-butyldimethylsilyl derivatization.
Determination of nitrogen mustard hydrolysis products, ethanolamines by gas chromatography-mass spectrometry after tert-butyldimethylsilyl derivatization.
Journal of chromatography. A (2006-05-19)
Isaac Ohsawa, Yasuo Seto
ABSTRACT
A method for determining N-ethyldiethanolamine (EDEA), N-methyldiethanolamine (MDEA) and triethanolamine (TEA), hydrolysis products of nitrogen mustards, in water, urine and blood samples using gas chromatography-mass spectrometry (GC-MS) after derivatization by tert-butyldimethylsilylation (TBDMS) is described. The sample solution was evaporated to dryness, and reacted with N-methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA) at 60 degrees C for 1h. The TBDMS derivatives were separated on a DB-5 column and detected by electron-ionization MS. The quantitation of EDEA, MDEA and TEA was performed by measuring the respective peak areas on the extracted ion chromatograms of m/z 216, m/z 202 and m/z 346, respectively, using nonadecane (C19), the peak area of which was measured at m/z 268, as an internal standard. When the water sample was initially analyzed, considerable loss of EDEA, MDEA and TEA occurred by evaporation. The addition of hydrochloric acid (HCl) to the water sample (final 1 mM), however, permitted quantitative recoveries to be achieved (88%, 88% and 79% for EDEA-(TBDMS)2, MDEA-(TBDMS)2 and TEA-(TBDMS)3, respectively). The limits of detections (LODs, scan mode, S/N = 3) were 2.5, 2.5 and 10 ng/ml for EDEA, MDEA and TEA, respectively. Ethanolamines could be also determined in urine samples (volume 0.1 ml), with reasonable recoveries of 72-100% by the addition of HCl (final 1 mM). For the analysis of serum samples, the sample was precipitated by the addition of perchloric acid (final 3.2%), and the resulting supernatant was neutralized with potassium carbonate, and then acidified by the addition of HCl. The recovery of TBDMS derivatives of ethanolamines was found to rather low (7-31%).
Product Description
Supelco
MTBSTFA (with 1% t-BDMCS), (with 1% t-BDMCS), ampule of 5 × 1 mL, analytical standard, Cerilliant®
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%