Merck
  • A study of TiO2/carbon black composition as counter electrode materials for dye-sensitized solar cells.
A study of TiO2/carbon black composition as counter electrode materials for dye-sensitized solar cells.
Nanoscale research letters (2013-05-16)
Jeongmin Lim, Sang Yeoul Ryu, Jeonghun Kim, Yongseok Jun
ABSTRACT
This study describes a systematic approach of TiO2/carbon black nanoparticles with respect to the loading amount in order to optimize the catalytic ability of triiodide reduction for dye-sensitized solar cells. In particular, the cell using an optimized TiO2 and carbon black electrode presents an energy conversion efficiency of 7.4% with a 5:1 ratio of a 40-nm TiO2 to carbon black. Based on the electrochemical analysis, the charge-transfer resistance of the carbon counter electrode changed based on the carbon black powder content. Electrochemical impedance spectroscopy and cyclic voltammetry study show lower resistance compared to the Pt counter electrode. The obtained nanostructures and photo electrochemical study were characterized.
Product Description
Supelco
Small Particle GCB Adsorbent, matrix Graphitized Carbon, <200 nm, bottle of 50 g
Pricing and availability is not currently available.