Merck
  • Mild manganese(III) acetate catalyzed allylic oxidation: application to simple and complex alkenes.
Mild manganese(III) acetate catalyzed allylic oxidation: application to simple and complex alkenes.
Organic letters (2006-06-30)
Tony K M Shing, Ying-Yeung Yeung, Pak L Su
ABSTRACT
Manganese(III) acetate catalyzed allylic oxidation of alkenes to the corresponding enones was investigated, showing excellent regioselectivity and chemoselectivity (functional group compatibility). Delta(5)-Steroids were transformed into bioactive Delta(5)-en-7-ones under a nitrogen atmosphere, whereas simple alkenes were converted into the corresponding enones under an oxygen atmosphere in good yields.
Sigma-Aldrich
Manganese(III) acetate dihydrate, 97%