Merck
  • Urinary phthalate metabolites in relation to preterm birth in Mexico city.
Urinary phthalate metabolites in relation to preterm birth in Mexico city.
Environmental health perspectives (2009-12-19)
John D Meeker, Howard Hu, David E Cantonwine, Hector Lamadrid-Figueroa, Antonia M Calafat, Adrienne S Ettinger, Mauricio Hernandez-Avila, Rita Loch-Caruso, Martha María Téllez-Rojo
ABSTRACT
Rates of preterm birth have been rising over the past several decades. Factors contributing to this trend remain largely unclear, and exposure to environmental contaminants may play a role. We investigated the relationship between phthalate exposure and preterm birth. Within a large Mexican birth cohort study, we compared third-trimester urinary phthalate metabolite concentrations in 30 women who delivered preterm (< 37 weeks of gestation) with those of 30 controls (> or = 37 weeks of gestation). Concentrations of most of the metabolites were similar to those reported among U.S. females, although in the present study mono-n-butyl phthalate (MBP) concentrations were higher and monobenzyl phthalate (MBzP) concentrations lower. In a crude comparison before correcting for urinary dilution, geometric mean urinary concentrations were higher for the phthalate metabolites MBP, MBzP, mono(3-carboxylpropyl) phthalate, and four metabolites of di(2-ethyl-hexyl) phthalate among women who subsequently delivered preterm. These differences remained, but were somewhat lessened, after correction by specific gravity or creatinine. In multivariate logistic regression analysis adjusted for potential confounders, elevated odds of having phthalate metabolite concentrations above the median level were found. We found that phthalate exposure is prevalent among this group of pregnant women in Mexico and that some phthalates may be associated with preterm birth.
Product Description
Supelco
mono-Benzyl phthalate, analytical standard
Pricing and availability is not currently available.