Merck
  • Macromolecular crowding directs extracellular matrix organization and mesenchymal stem cell behavior.
Macromolecular crowding directs extracellular matrix organization and mesenchymal stem cell behavior.
PloS one (2012-06-01)
Adam S Zeiger, Felicia C Loe, Ran Li, Michael Raghunath, Krystyn J Van Vliet
ABSTRACT
Microenvironments of biological cells are dominated in vivo by macromolecular crowding and resultant excluded volume effects. This feature is absent in dilute in vitro cell culture. Here, we induced macromolecular crowding in vitro by using synthetic macromolecular globules of nm-scale radius at physiological levels of fractional volume occupancy. We quantified the impact of induced crowding on the extracellular and intracellular protein organization of human mesenchymal stem cells (MSCs) via immunocytochemistry, atomic force microscopy (AFM), and AFM-enabled nanoindentation. Macromolecular crowding in extracellular culture media directly induced supramolecular assembly and alignment of extracellular matrix proteins deposited by cells, which in turn increased alignment of the intracellular actin cytoskeleton. The resulting cell-matrix reciprocity further affected adhesion, proliferation, and migration behavior of MSCs. Macromolecular crowding can thus aid the design of more physiologically relevant in vitro studies and devices for MSCs and other cells, by increasing the fidelity between materials synthesized by cells in vivo and in vitro.
Sigma-Aldrich
Poly(ethylene glycol), BioUltra, 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Pricing and availability is not currently available.