Merck
  • The zinc finger protein Zfr1p is localized specifically to conjugation junction and required for sexual development in Tetrahymena thermophila.
The zinc finger protein Zfr1p is localized specifically to conjugation junction and required for sexual development in Tetrahymena thermophila.
PloS one (2012-12-20)
Jing Xu, Huaru Tian, Wei Wang, Aihua Liang
ABSTRACT
Conjugation in Tetrahymena thermophila involves a developmental program consisting of three prezygotic nuclear divisions, pronuclear exchange and fusion, and postzygotic and exconjugant stages. The conjugation junction structure appears during the initiation of conjugation development, and disappears during the exconjugant stage. Many structural and functional proteins are involved in the establishment and maintenance of the junction structure in T. thermophila. In the present study, a zinc finger protein-encoding gene ZFR1 was found to be expressed specifically during conjugation and to localize specifically to the conjugation junction region. Truncated Zfr1p localized at the plasma membrane in ordered arrays and decorated Golgi apparatus located adjacent to basal body. The N-terminal zinc finger and C-terminal hydrophobic domains of Zfr1p were found to be required for its specific conjugation junction localization. Conjugation development of ZFR1 somatic knockout cells was aborted at the pronuclear exchange and fusion conjugation stages. Furthermore, Zfr1p was found to be important for conjugation junction stability during the prezygotic nuclear division stage. Taken together, our data reveal that Zfr1p is required for the stability and integrity of the conjugation junction structure and essential for the sexual life cycle of the Tetrahymena cell.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, F(ab′)2, FITC conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Anti-HA Tag Antibody, clone 114-2C-7, rabbit monoclonal, clone 114-2C-7, Upstate®, from rabbit