Merck
  • Duration differences of corticostriatal responses in striatal projection neurons depend on calcium activated potassium currents.
Duration differences of corticostriatal responses in striatal projection neurons depend on calcium activated potassium currents.
Frontiers in systems neuroscience (2013-10-11)
Mario A Arias-García, Dagoberto Tapia, Edén Flores-Barrera, Jesús E Pérez-Ortega, José Bargas, Elvira Galarraga
ABSTRACT
The firing of striatal projection neurons (SPNs) exhibits afterhyperpolarizing potentials (AHPs) that determine discharge frequency. They are in part generated by Ca(2+)-activated K(+)-currents involving BK and SK components. It has previously been shown that suprathreshold corticostriatal responses are more prolonged and evoke more action potentials in direct pathway SPNs (dSPNs) than in indirect pathway SPNs (iSPNs). In contrast, iSPNs generate dendritic autoregenerative responses. Using whole cell recordings in brain slices, we asked whether the participation of Ca(2+)-activated K(+)-currents plays a role in these responses. Secondly, we asked if these currents may explain some differences in synaptic integration between dSPNs and iSPNs. Neurons obtained from BAC D1 and D2 GFP mice were recorded. We used charybdotoxin and apamin to block BK and SK channels, respectively. Both antagonists increased the depolarization and delayed the repolarization of suprathreshold corticostriatal responses in both neuron classes. We also used NS 1619 and NS 309 (CyPPA), to enhance BK and SK channels, respectively. Current enhancers hyperpolarized and accelerated the repolarization of corticostriatal responses in both neuron classes. Nevertheless, these drugs made evident that the contribution of Ca(2+)-activated K(+)-currents was different in dSPNs as compared to iSPNs: in dSPNs their activation was slower as though calcium took a diffusion delay to activate them. In contrast, their activation was fast and then sustained in iSPNs as though calcium flux activates them at the moment of entry. The blockade of Ca(2+)-activated K(+)-currents made iSPNs to look as dSPNs. Conversely, their enhancement made dSPNs to look as iSPNs. It is concluded that Ca(2+)-activated K(+)-currents are a main intrinsic determinant causing the differences in synaptic integration between corticostriatal polysynaptic responses between dSPNs and iSPNs.
Sigma-Aldrich
CyPPA, ≥98% (HPLC), solid
Sigma-Aldrich
NS309, ≥98% (HPLC), solid
Sigma-Aldrich
NS 1619, powder