Merck
  • Systemic PPARγ deletion impairs circadian rhythms of behavior and metabolism.
Systemic PPARγ deletion impairs circadian rhythms of behavior and metabolism.
PloS one (2012-08-18)
Guangrui Yang, Zhanjun Jia, Toshinori Aoyagi, Donald McClain, Richard M Mortensen, Tianxin Yang
ABSTRACT
Compelling evidence from both human and animal studies suggests a physiological link between the circadian rhythm and metabolism but the underlying mechanism is still incompletely understood. We examined the role of PPARγ, a key regulator of energy metabolism, in the control of physiological and behavioral rhythms by analyzing two strains of whole-body PPARγ null mouse models. Systemic inactivation of PPARγ was generated constitutively by using Mox2-Cre mice (MoxCre/flox) or inducibly by using the tamoxifen system (EsrCre/flox/TM). Circadian variations in oxygen consumption, CO(2) production, food and water intake, locomotor activity, and cardiovascular parameters were all remarkably suppressed in MoxCre/flox mice. A similar phenotype was observed in EsrCre/flox/TM mice, accompanied by impaired rhythmicity of the canonical clock genes in adipose tissues and liver but not skeletal muscles or the kidney. PPARγ inactivation in isolated preadipocytes following exposure to tamoxifen led to a similar blockade of the rhythmicity of the clock gene expression. Together, these results support an essential role of PPARγ in the coordinated control of circadian clocks and metabolic pathways.
Sigma-Aldrich
Krebs-Ringer Bicarbonate Buffer, With 1800 mg/L glucose, without calcium chloride and sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Pricing and availability is not currently available.
Sigma-Aldrich
Tamoxifen, ≥99%
Pricing and availability is not currently available.