Merck
  • Insulin-like growth factor-I E-peptide activity is dependent on the IGF-I receptor.
Insulin-like growth factor-I E-peptide activity is dependent on the IGF-I receptor.
PloS one (2012-10-03)
Becky K Brisson, Elisabeth R Barton
ABSTRACT
Insulin-like growth factor-I (IGF-I) is an essential growth factor that regulates the processes necessary for cell proliferation, differentiation, and survival. The Igf1 gene encodes mature IGF-I and a carboxy-terminal extension called the E-peptide. In rodents, alternative splicing and post-translational processing produce two E-peptides (EA and EB). EB has been studied extensively and has been reported to promote cell proliferation and migration independently of IGF-I and its receptor (IGF-IR), but the mechanism by which EB causes these actions has not been identified. Further, the properties of EA have not been evaluated. Therefore, the goals of this study were to determine if EA and EB possessed similar activity and if these actions were IGF-IR independent. We utilized synthetic peptides for EA, EB, and a scrambled control to examine cellular responses. Both E-peptides increased MAPK signaling, which was blocked by pharmacologic IGF-IR inhibition. Although the E-peptides did not directly induce IGF-IR phosphorylation, the presence of either E-peptide increased IGF-IR activation by IGF-I, and this was achieved through enhanced cell surface bioavailability of the receptor. To determine if E-peptide biological actions required the IGF-IR, we took advantage of the murine C2C12 cell line as a platform to examine the key steps of skeletal muscle proliferation, migration and differentiation. EB increased myoblast proliferation and migration while EA delayed differentiation. The proliferation and migration effects were inhibited by MAPK or IGF-IR signaling blockade. Thus, in contrast to previous studies, we find that E-peptide signaling, mitogenic, and motogenic effects are dependent upon IGF-IR. We propose that the E-peptides have little independent activity, but instead affect growth via modulating IGF-I signaling, thereby increasing the complexity of IGF-I biological activity.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-Insulin-like Growth Factor-1 Receptor Antibody, α-subunit, clone 24-31, clone 24-31, Chemicon®, from mouse
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone PY20, HRP Conjugate, clone PY20, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2