Merck
  • Recent highlights in electrophilic fluorination with 1-chloromethyl-4-fluoro- 1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate).
Recent highlights in electrophilic fluorination with 1-chloromethyl-4-fluoro- 1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate).
Accounts of chemical research (2004-01-21)
Rajendra P Singh, Jean'ne M Shreeve
ABSTRACT
Synthetic and structural aspects of organofluorine compounds continue to be the focal points of vigorous research activities, as evidenced by the appearance of a large number of publications. Among the various useful methodologies for the introduction of fluorine into organic molecules, electrophilic fluorination is a promising and exciting area of research. While a variety of electrophilic fluorinating reagents are available, currently 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) (Selectfluor) provides a remarkably straightforward and effective route. The breadth of applications realizable from this reagent in its role as a key electrophilic fluorinating reagent is highlighted here. This Account covers the literature for electrophilic fluorination reactions that employ Selectfluor during the period January 1999-January 2003.
Sigma-Aldrich
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), >95% in F+ active
Pricing and availability is not currently available.