Merck
  • Practical syntheses of N-substituted 3-hydroxyazetidines and 4-hydroxypiperidines by hydroxylation with Sphingomonas sp. HXN-200.
Practical syntheses of N-substituted 3-hydroxyazetidines and 4-hydroxypiperidines by hydroxylation with Sphingomonas sp. HXN-200.
Organic letters (2002-05-25)
Dongliang Chang, Hans-Jürgen Feiten, Karl-H Engesser, Jan B van Beilen, Bernard Witholt, Zhi Li
ABSTRACT
[reaction: see text] Hydroxylation of N-substituted azetidines 11 and 12 and piperidines 15-19 with Sphingomonas sp. HXN-200 gave 91-98% of the corresponding 3-hydroxyazetidines 13 and 14 and 4-hydroxypiperidines 20-24, respectively, with high activity and excellent regioselectivity. High yields and high product concentrations (2 g/L) were achieved with frozen/thawed cells as biocatalyst. For the first time, rehydrated lyophilized cells were successfully used for the biohydroxylation.
Sigma-Aldrich
Azetidine hydrochloride, 97%