Merck
  • Effect of Steric Constraint at the γ-Backbone Position on the Conformations and Hybridization Properties of PNAs.
Effect of Steric Constraint at the γ-Backbone Position on the Conformations and Hybridization Properties of PNAs.
Journal of nucleic acids (2011-07-22)
Matthew J Crawford, Srinivas Rapireddy, Raman Bahal, Iulia Sacui, Danith H Ly
ABSTRACT
Conformationally preorganized peptide nucleic acids (PNAs) have been synthesized through backbone modifications at the γ-position, where R = alanine, valine, isoleucine, and phenylalanine side chains. The effects of these side-chains on the conformations and hybridization properties of PNAs were determined using a combination of CD and UV-Vis spectroscopic techniques. Our results show that the γ-position can accommodate varying degrees of sterically hindered side-chains, reaffirming the bimodal function of PNAs as the true hybrids of "peptides" and "nucleic acids."
Sigma-Aldrich
Boc-Phe-OH, ≥99.0% (T)
Pricing and availability is not currently available.