Merck
  • Water-soluble renin inhibitors: design of a subnanomolar inhibitor with a prolonged duration of action.
Water-soluble renin inhibitors: design of a subnanomolar inhibitor with a prolonged duration of action.
Journal of medicinal chemistry (1990-07-01)
S H Rosenberg, K W Woods, H L Sham, H D Kleinert, D L Martin, H Stein, J Cohen, D A Egan, B Bopp, I Merits
ABSTRACT
Incorporation of nonreactive polar functionalities at the C- and N-termini of renin inhibitors led to the development of a subnanomolar compound (21) with millimolar solubility. This inhibitor demonstrated excellent efficacy and a long duration of action upon intravenous administration to monkeys. While activity was also observed intraduodenally, a comparison of the blood pressure responses indicated low bioavailability. Subsequent experiments in rats showed that, although the compound was absorbed from the gastrointestinal tract, extensive liver extraction severely limited bioavailability.
Sigma-Aldrich
3-Pyrroline, 95%
Pricing and availability is not currently available.