Merck
  • Evolutionary breakpoints in the gibbon suggest association between cytosine methylation and karyotype evolution.
Evolutionary breakpoints in the gibbon suggest association between cytosine methylation and karyotype evolution.
PLoS genetics (2009-06-27)
Lucia Carbone, R Alan Harris, Gery M Vessere, Alan R Mootnick, Sean Humphray, Jane Rogers, Sung K Kim, Jeffrey D Wall, David Martin, Jerzy Jurka, Aleksandar Milosavljevic, Pieter J de Jong
ABSTRACT
Gibbon species have accumulated an unusually high number of chromosomal changes since diverging from the common hominoid ancestor 15-18 million years ago. The cause of this increased rate of chromosomal rearrangements is not known, nor is it known if genome architecture has a role. To address this question, we analyzed sequences spanning 57 breaks of synteny between northern white-cheeked gibbons (Nomascus l. leucogenys) and humans. We find that the breakpoint regions are enriched in segmental duplications and repeats, with Alu elements being the most abundant. Alus located near the gibbon breakpoints (<150 bp) have a higher CpG content than other Alus. Bisulphite allelic sequencing reveals that these gibbon Alus have a lower average density of methylated cytosine that their human orthologues. The finding of higher CpG content and lower average CpG methylation suggests that the gibbon Alu elements are epigenetically distinct from their human orthologues. The association between undermethylation and chromosomal rearrangement in gibbons suggests a correlation between epigenetic state and structural genome variation in evolution.
Product Description
Roche
FastStart Taq DNA Polymerase, 5 U/μl, dNTPs included: no, hotstart, suitable for PCR