Merck
  • Lipoxygenation of arachidonic acid as a source of polymorphonuclear leukocyte chemotactic factors in synovial fluid and tissue in rheumatoid arthritis and spondyloarthritis.
Lipoxygenation of arachidonic acid as a source of polymorphonuclear leukocyte chemotactic factors in synovial fluid and tissue in rheumatoid arthritis and spondyloarthritis.
The Journal of clinical investigation (1980-11-01)
L B Klickstein, C Shapleigh, E J Goetzl
ABSTRACT
The predominant lipoxygenase products of arachidonic acid were extracted and purified from synovial fluid and sonicates of synovial tissue of patients with rheumatoid arthritis (RA), spondyloarthritis (SA), or a noninflammatory arthropathy (NIA). The concentration of 5(S),12(R)-dihydroxy-6,8,10-(trans/trans/cis)-14-cis-eicosatetraenoic acid (leukotriene B4) in synovial fluid was elevated significantly in patients with RA and a positive latex test for rheumatoid factor (P < 0.05, n = 14) and in patients with SA (P < 0.05, n = 10), compared with that of subjects with NIA (n = 9). The content of 5(S)-hydroxy-6,8,11,14-eicosatetraenoic acid (5-HETE), but not of leukotriene B4, was elevated significantly in synovial tissue of seven patients with RA in comparison with that of four subjects with NIA (P < 0.05). A single intra-articular injection of corticosteroid significantly lowered the synovial fluid level of leukotriene B4 in six patients with RA. These data suggest an involvement of the potent chemotactic factors 5-HETE and leukotriene B4 in human inflammatory disease.
Sigma-Aldrich
Arachidonic acid sodium salt from Mortierella alpina, ≥98.5% (GC)