Merck
  • The effect of glucagon-like peptide 2 injection on performance, small intestinal morphology, and nutrient transporter expression of stressed broiler chickens.
The effect of glucagon-like peptide 2 injection on performance, small intestinal morphology, and nutrient transporter expression of stressed broiler chickens.
Poultry science (2010-08-17)
X F Hu, Y M Guo, B Y Huang, S Bun, L B Zhang, J H Li, D Liu, F Y Long, X Yang, P Jiao
ABSTRACT
An experiment was conducted to determine the effect of injecting glucagon-like peptide 2 (GLP-2) on the small intestinal weight, morphology, and nutrient transporter expression in pharmacologically stressed broiler chickens. A total of 144 seven-day-old birds were fed either a basal diet (CTRL) or a basal diet plus 30 mg of corticosterone (CORT)/kg of diet for a total of 14 d. Half of the birds from each group were injected daily with GLP-2 (6.7 nmol/kg of BW) or saline for 14 d. The average final BW, ADG, ADFI, and the ratio of feed intake to weight gain (F:G) was recorded over 21 d for the 4 groups of 36 birds, namely CTRL + saline, CTRL + GLP-2, CORT + saline, and CORT + GLP-2. In addition, the absolute and relative small intestinal weight, villus height (VH), and crypt depth (CD) of the duodenum and jejunum, as well as the abundance of sodium and glucose co-transporter 1 (SGLT-1), vitamin D-dependent calcium-binding protein-28,000 molecular weight (CaBP-D28k), and peptide transporter 1 (PepT-1) mRNA in the duodenum and of liver fatty acid-binding protein (L-FABP) mRNA in the jejunum. The total DNA, RNA, and protein content in small intestinal mucosa were also determined. The results showed that CORT administration significantly lowered average final BW, ADG, ADFI, absolute small intestinal weight, VH, and CD of duodenum and jejunum (P < 0.05) while increasing the relative small intestinal weight, F:G, relative abundance of SGLT-1, CaBP-D28k, PepT-1, and L-FABP mRNA (P < 0.05). Glucagon-like peptide 2 injection increased the average final BW, ADG, VH, and CD in duodenum and jejunum and relative abundance of SGLT-1, CaBP-28k, PepT1, and PepT1 mRNA of broiler chickens, respectively (P < 0.05), and decreased F:G (P < 0.05). In chickens fed basal diet plus CORT, injecting GLP-2 decreased F:G (P < 0.05); increased VH and CD of duodenum and CD of jejunum; and increased relative abundance of SGLT-1, CaBP-D28k, PepT-1, and L-FABP mRNA, RNA, and total protein content in small intestine compared with the injection of saline (P < 0.05). In birds fed the basal diet, GLP-2 injection decreased F:G (P < 0.05) and increased final BW, ADG, small bowel weight, CD of jejunum, and relative abundance of CaBP-D28k and PepT-1 mRNA compared with injecting saline (P < 0.05). In conclusion, GLP-2 injection reversed the negative effect of stress on the weight and morphology and the absorptive function of small bowel of broiler chickens. Glucagon-like peptide 2 injection also had a positive effect on the growth performance of healthy broiler chickens.
Sigma-Aldrich
Glucagon-Like Peptide 2 -Arg human trifluoroacetate salt, >90% (HPLC), solid
Pricing and availability is not currently available.