Merck
  • Neutrophils Self-Regulate Immune Complex-Mediated Cutaneous Inflammation through CXCL2.
Neutrophils Self-Regulate Immune Complex-Mediated Cutaneous Inflammation through CXCL2.
The Journal of investigative dermatology (2016-01-24)
Jackson LiangYao Li, Chun Hwee Lim, Fen Wei Tay, Chi Ching Goh, Sapna Devi, Benoit Malleret, Bernett Lee, Nadja Bakocevic, Shu Zhen Chong, Maximilien Evrard, Hideaki Tanizaki, Hwee Ying Lim, Bruce Russell, Laurent Renia, Francesca Zolezzi, Michael Poidinger, Veronique Angeli, Ashley L St John, John E Harris, Hong Liang Tey, Suet Mien Tan, Kenji Kabashima, Wolfgang Weninger, Anis Larbi, Lai Guan Ng
ABSTRACT
Deposition of immune complexes (ICs) in tissues triggers acute inflammatory pathology characterized by massive neutrophil influx leading to edema and hemorrhage, and is especially associated with vasculitis of the skin, but the mechanisms that regulate this type III hypersensitivity process remain poorly understood. Here, using a combination of multiphoton intravital microscopy and genomic approaches, we re-examined the cutaneous reverse passive Arthus reaction and observed that IC-activated neutrophils underwent transmigration, triggered further IC formation, and transported these ICs into the interstitium, whereas neutrophil depletion drastically reduced IC formation and ameliorated vascular leakage in vivo. Thereafter, we show that these neutrophils expressed high levels of CXCL2, which further amplified neutrophil recruitment and activation in an autocrine and/or paracrine manner. Notably, CXCL1 expression was restricted to tissue-resident cell types, but IC-activated neutrophils may also indirectly, via soluble factors, modulate macrophage CXCL1 expression. Consistent with their distinct cellular origins and localization, only neutralization of CXCL2 but not CXCL1 in the interstitium effectively reduced neutrophil recruitment. In summary, our study establishes that neutrophils are able to self-regulate their own recruitment and responses during IC-mediated inflammation through a CXCL2-driven feed forward loop.
Sigma-Aldrich
Anti-Bovine Albumin antibody produced in rabbit, fractionated antiserum, lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture