Merck
  • Anti-Inflammatory Effects of β2-Receptor Agonists Salbutamol and Terbutaline Are Mediated by MKP-1.
Anti-Inflammatory Effects of β2-Receptor Agonists Salbutamol and Terbutaline Are Mediated by MKP-1.
PloS one (2016-02-06)
Tiina Keränen, Tuija Hömmö, Mari Hämäläinen, Eeva Moilanen, Riku Korhonen
ABSTRACT
Mitogen-activated protein kinase phosphatase 1 (MKP-1) expression is induced by inflammatory factors, and it is an endogenous suppressor of inflammatory response. MKP-1 expression is increased by PDE4 inhibitor rolipram suggesting that it is regulated by cAMP-enhancing compounds. Therefore, we investigated the effect of β2-receptor agonists on MKP-1 expression and inflammatory response. We found that β2-receptor agonists salbutamol and terbutaline, as well as 8-Br-cAMP, increased MKP-1 expression. Salbutamol and terbutaline also inhibited p38 MAPK phosphorylation and TNF production in J774 mouse macrophages. Interestingly, salbutamol suppressed carrageenan-induced paw inflammation in wild-type mice, but the effect was attenuated in MKP-1(-/-) mice. In conclusion, these data show that β2-receptor agonists increase MKP-1 expression, which seems to mediate, at least partly, the observed anti-inflammatory effects of β2-receptor agonists.
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder