Merck
Faster evolution of highly conserved DNA in tropical plants.
Journal of evolutionary biology (2010-05-12)
Len N Gillman, D J Keeling, R C Gardner, S D Wright
ABSTRACT
A faster rate of nuclear DNA evolution has recently been found for plants occupying warmer low latitudes relative to those in cooler high latitudes. That earlier study by our research group compared substitution rates within the variable internal transcribed spacer (ITS) region of the ribosomal gene complex amongst 45 congeneric species pairs, each member of which differed in their latitudinal distributions. To determine whether this rate differential might also occur within highly conserved DNA, we sequenced the 18S ribosomal gene in the same 45 pairs of plants. We found that the rate of evolution in 18S was 51% faster in the tropical plant species relative to their temperate sisters and that the substitution rate in 18S correlated positively with that in the more variable ITS. This result, with a gene coding for ribosomal structure, suggests that climatic influences on evolution extend to functionally important regions of the genome.
Sigma-Aldrich
Extract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
Extract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 500 amplifications
Pricing and availability is not currently available.