Merck
  • Enhanced seed phytosterol accumulation through expression of a modified HMG-CoA reductase.
Enhanced seed phytosterol accumulation through expression of a modified HMG-CoA reductase.
Plant biotechnology journal (2006-12-21)
Sandra J Hey, Stephen J Powers, Michael H Beale, Nathaniel D Hawkins, Jane L Ward, Nigel G Halford
ABSTRACT
The regulation of phytosterol biosynthesis in seeds is of interest to biotechnologists because of the efficacy of dietary phytosterols in reducing blood cholesterol in humans. Mevalonate synthesis via 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMG-CoA reductase) is a key step in phytosterol biosynthesis. HMG-CoA reductase is inactivated by phosphorylation by SNF1-related protein kinase 1 (SnRK1). With the aim of increasing seed phytosterol levels, transgenic tobacco plants were produced expressing a full-length Arabidopsis (Arabidopsis thaliana) HMG-CoA reductase gene (HMG1) coding sequence, a modified HMG1 sequence encoding a protein lacking the target serine residue for phosphorylation by SnRK1, or a chimaeric sequence encoding the N-terminal domain of the Arabidopsis HMG1 enzyme fused with the catalytic domain of yeast HMG-CoA reductase, which lacks an SnRK1 target site. All three transgenes (35S-AtHMG1, 35S-AtHMG1m and 35S-AtScHMG1) were under the control of a cauliflower mosaic virus 35S RNA promoter. Levels of seed phytosterols were up to 2.44-fold higher in plants transformed with the 35S-AtHMG1m gene than in the wild-type, and were significantly higher than in plants expressing 35S-AtHMG1 or 35S-AtScHMG1. In contrast, levels of phytosterols in leaves of plants transformed with the 35S-AtHMG1m gene were unchanged, suggesting that regulation of HMG-CoA reductase by SnRK1 is an important factor in seeds but not in leaves. A total of 11 independent transgenic lines expressing 35S-AtHMG1m or 35S-AtScHMG1 also showed an altered flower phenotype, comprising a compact floret, prolonged flowering, short, pale petals, a protruding style, short stamens, late anther development, little or no pollen production, premature flower abscission and poor seed set. Because of this phenotype, the modified HMG-CoA reductase gene would have to be expressed seed specifically if it were to be engineered into a crop plant for biotechnological purposes.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Pricing and availability is not currently available.
Sigma-Aldrich
REDExtract-N-Amp Plant PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Pricing and availability is not currently available.