Merck
A role for Separase in telomere protection.
Nature communications (2016-01-19)
Francesca Cipressa, Patrizia Morciano, Giuseppe Bosso, Linda Mannini, Alessandra Galati, Grazia Daniela Raffa, Stefano Cacchione, Antonio Musio, Giovanni Cenci
ABSTRACT
Drosophila telomeres are elongated by transposition of specialized retroelements rather than telomerase activity and are assembled independently of the sequence. Fly telomeres are protected by the terminin complex that localizes and functions exclusively at telomeres and by non-terminin proteins that do not serve telomere-specific functions. We show that mutations in the Drosophila Separase encoding gene Sse lead not only to endoreduplication but also telomeric fusions (TFs), suggesting a role for Sse in telomere capping. We demonstrate that Separase binds terminin proteins and HP1, and that it is enriched at telomeres. Furthermore, we show that loss of Sse strongly reduces HP1 levels, and that HP1 overexpression in Sse mutants suppresses TFs, suggesting that TFs are caused by a HP1 diminution. Finally, we find that siRNA-induced depletion of ESPL1, the Sse human orthologue, causes telomere dysfunction and HP1 level reduction in primary fibroblasts, highlighting a conserved role of Separase in telomere protection.
Sigma-Aldrich
Anti-53BP1 Antibody, clone BP18, ascites fluid, clone BP18, Upstate®
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Anti-Digoxigenin-Rhodamine, Fab fragments, from sheep
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1