Merck
  • CD163 interacts with TWEAK to regulate tissue regeneration after ischaemic injury.
CD163 interacts with TWEAK to regulate tissue regeneration after ischaemic injury.
Nature communications (2015-08-06)
Hirokuni Akahori, Vinit Karmali, Rohini Polavarapu, Alicia N Lyle, Daiana Weiss, Eric Shin, Ahsan Husain, Nawazish Naqvi, Richard Van Dam, Anwer Habib, Cheol Ung Choi, Adrienne L King, Kimberly Pachura, W Robert Taylor, David J Lefer, Aloke V Finn
ABSTRACT
Macrophages are an essential component of the immune response to ischaemic injury and play an important role in promoting inflammation and its resolution, which is necessary for tissue repair. The type I transmembrane glycoprotein CD163 is exclusively expressed on macrophages, where it acts as a receptor for haemoglobin:haptoglobin complexes. An extracellular portion of CD163 circulates in the blood as a soluble protein, for which no physiological function has so far been described. Here we show that during ischaemia, soluble CD163 functions as a decoy receptor for TWEAK, a secreted pro-inflammatory cytokine of the tumour necrosis factor family, to regulate TWEAK-induced activation of canonical nuclear factor-κB (NF-κB) and Notch signalling necessary for myogenic progenitor cell proliferation. Mice with deletion of CD163 have transiently elevated levels of TWEAK, which stimulate muscle satellite cell proliferation and tissue regeneration in their ischaemic and non-ischaemic limbs. These results reveal a role for soluble CD163 in regulating muscle regeneration after ischaemic injury.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), FITC conjugate, lyophilized powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Syt1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rbpj
Sigma-Aldrich
MISSION® esiRNA, targeting human SYT1