Merck
  • Determination of Urinary PAH Metabolites Using DLLME Hyphenated to Injector Port Silylation and GC-MS-MS.
Determination of Urinary PAH Metabolites Using DLLME Hyphenated to Injector Port Silylation and GC-MS-MS.
Journal of analytical toxicology (2015-04-08)
Manoj Kumar Gupta, Rajeev Jain, Pratibha Singh, Ratnasekhar Ch, Mohana Krishna Reddy Mudiam
ABSTRACT
Polycyclic aromatic hydrocarbons (PAHs) are ubiquitous environmental pollutants and well-known carcinogens. Hydroxy derivatives of PAH are considered as biomarkers of PAH exposure, and there is a need to measure these metabolites at low concentrations. So, a precise and eco-friendly analytical method has been developed for rapid determination of PAH metabolites. For the first time, a new analytical method based on coupling of dispersive liquid-liquid microextraction (DLLME) with auto-injector port silylation (auto-IPS) followed by gas chromatography-tandem mass spectrometry (GC-MS-MS) analysis is reported for the analysis of seven urinary PAH metabolites. Factors affecting DLLME and IPS, such as type and volume of extraction and disperser solvent, pH, ionic strength, injector port temperature, volume of N,O-bis(trimethylsilyl)trifluoroacetamide and type of solvent were investigated. Under optimized conditions, the limit of detection and limit of quantification were found to be in the range of 1-9 and 3-29 ng/mL, respectively. Satisfactory recoveries of metabolites in urine samples in the range of 87-95% were found. The developed method has been successfully applied for the determination of PAH metabolites in urine samples of exposed workers. DLLME-auto-IPS-GC-MS-MS method is time, labor, solvent and reagent saving, which can be routinely used for the analysis of urinary PAH metabolites.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Tetrachloroethylene, anhydrous, ≥99%
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hexarelin, ≥90% (HPLC)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Ultrapure Acetonitrile