Merck
  • A Novel Electrochemiluminescence Immunosensor for the Analysis of HIV-1 p24 Antigen Based on P-RGO@Au@Ru-SiO₂ Composite.
A Novel Electrochemiluminescence Immunosensor for the Analysis of HIV-1 p24 Antigen Based on P-RGO@Au@Ru-SiO₂ Composite.
ACS applied materials & interfaces (2015-10-22)
Limin Zhou, Jianshe Huang, Bin Yu, Yang Liu, Tianyan You
ABSTRACT
Ru(bpy)3(2+)-doped silica (Ru-SiO2) nanoparticles and gold-nanoparticle-decorated graphene (P-RGO@Au) were combined to form a P-RGO@Au@Ru-SiO2 composite. The composite was used to develop a novel sandwich-type electrochemiluminescence immunosensor for the analysis of HIV-1 p24 antigen. The composite worked as carrier to immobilize target antibody and to build a sandwich-type electrochemiluminescence immunosensor through an interaction between antigen and antibody. Importantly, high ECL signal could be obtained due to the large amounts of Ru(bpy)3(2+) molecules per Ru-SiO2 nanoparticle. The P-RGO@Au composite with good conductivity and high surface area not only accelerated the electron transfer rate but also improved the loading of both ECL molecules and capture antibody, which could further increase the ECL response and result in high sensitivity. Taking advantage of both Ru-SiO2 nanoparticles and the P-RGO@Au composite, the proposed immunosensor exhibited a linear range from 1.0 × 10(-9) to 1.0 × 10(-5) mg mL(-1) with a detection limit of 1.0 × 10(-9) mg mL(-1) for HIV-1 p24 antigen. The proposed ECL immunosensor was used to analyze HIV-1 p24 antigen in human serum, and satisfactory recoveries were obtained, indicating that the proposed method is promising for practical applications in the clinical diagnosis of HIV infection.
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Tripropylamine, ≥98%
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Hexanol, reagent grade, 98%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexyl alcohol, FCC, FG
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)