Merck
  • Effects of Dietary L-carnosine and Alpha-lipoic Acid on Growth Performance, Blood Thyroid Hormones and Lipid Profiles in Finishing Pigs.
Effects of Dietary L-carnosine and Alpha-lipoic Acid on Growth Performance, Blood Thyroid Hormones and Lipid Profiles in Finishing Pigs.
Asian-Australasian journal of animal sciences (2015-07-22)
Yinghui Bao, Chunqi Gao, Wenbo Hao, Cheng Ji, Lihong Zhao, Jianyun Zhang, Tao Liu, Qiugang Ma
ABSTRACT
The present study was conducted to determine the effects of L-carnosine (LC) and/or alpha-lipoic acid (ALA) supplementation on growth performance, blood thyroid hormones and lipid profiles in finishing pigs. A total of 40 (Landrace×Yorkshire) pigs with an initial body weight of 57.93±3.14 kg were randomly allocated to 4 experimental diets using a 2×2 factorial arrangement with 2 LC supplemental levels (0 or 0.1%) and 2 ALA supplemental levels (0 or 0.03%) in basal diets. The results showed that pigs fed LC-supplemented diets increased final live weight, average daily gain, and average daily feed intake compared to those of pigs fed without LC-supplemented diets (p<0.05). Dietary supplementation with ALA did not affect the growth performance and carcass traits of pigs (p>0.05). Additionally, LC supplementation increased serum triiodothyronine, thyroxine levels, and ALA supplementation increased serum triiodothyronine levels (p<0.05). Serum total cholesterol and triglycerides levels were significantly decreased in LC and ALA supplemented groups, respectively (p<0.05). Moreover, serum low density lipoprotein cholesterol levels were lower in the ALA-supplemented groups than those of pigs fed without ALA-supplemented diets (p<0.05). However, no significant LC×ALA interaction effect on growth performance, blood thyroid hormones and lipid profiles was found. This study suggested that dietary supplementation of LC resulted in better growth performance compared to that of ALA supplementation. L-carnosine and/or ALA supplementation positively modified blood lipid profiles, which may have the potential to prevent cardiovascular diseases.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®