Merck
  • Benzophenone 1 induced photogenotoxicity and apoptosis via release of cytochrome c and Smac/DIABLO at environmental UV radiation.
Benzophenone 1 induced photogenotoxicity and apoptosis via release of cytochrome c and Smac/DIABLO at environmental UV radiation.
Toxicology letters (2015-10-07)
Saroj Kumar Amar, Shruti Goyal, Divya Dubey, Ajeet K Srivastav, Deepti Chopra, Jyoti Singh, Jai Shankar, Rajnish K Chaturvedi, Ratan Singh Ray
ABSTRACT
Solar UV radiation is main factor of photocarcinogenesis, photoageing, and phototoxicity; thus, protection from UV radiation is major concern. Sunscreens containing UV filters are suggested as sun safe practices, but safety of UV filters remains in controversies. Benzophenone-1 (BP1) is commonly used in sunscreens as UV blocker. We assessed the photogenotoxicity and apoptotic parameters in human keratinocytes (HaCaT cells) by western blot, immunocytochemistry, flowcytometry, comet assay and TEM imaging. Our results exposed that BP1 photosensitized and generated intracellular ROS (2.02 folds) under sunlight/UVR. Decrease in cell viability was recorded as 80.06%, 60.98% and 56.24% under sunlight, UVA and UVB, respectively. Genotoxic potential of BP1 was confirmed through photomicronuclei and CPDs formation. BP1 enhanced lipid peroxidation and leakage of LDH enzyme (61.7%). Apoptotic cells were detected by AnnexinV/PI staining and sub G1 population of cell cycle. BP1 induced up regulation of apoptotic proteins Bax/Bcl2 ratio, Apaf-1, cytochrome c, Smac/DIABLO and cleaved caspase 3 was noticed. Down regulation of pro caspase 3 was inhibited by Z-VAD-fmk (inhibitor of caspase). Thus, study established the involvement of BP1 in photogenotoxicity and apoptosis via release of cytochrome c and Smac/DIABLO. These findings suggest sunscreen user to avoid BP1 in cosmetics preparation for its topical application.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%