Merck
  • Abnormal N-acetylglucosaminyltransferase expression in prefrontal cortex in schizophrenia.
Abnormal N-acetylglucosaminyltransferase expression in prefrontal cortex in schizophrenia.
Schizophrenia research (2015-06-25)
Jordyn M Kippe, Toni M Mueller, Vahram Haroutunian, James H Meador-Woodruff
ABSTRACT
Changes in the extent of the posttranslational modification glycosylation have been previously reported in several brain regions in schizophrenia. Quality control within the endoplasmic reticulum and Golgi, branching of glycans, intracellular trafficking and targeting, protein-protein interactions, and endocytosis are processes regulated by both N-linked and O-linked glycosylation. Previous studies in schizophrenia have found altered glycan biosynthesis and abnormal glycan levels in cerebrospinal fluid (CSF) and plasma, as well as altered expression in frontal cortex of glycosyltransferase transcripts encoding proteins associated with both N- and O-linked glycosylation. The N-acetylglucosaminyltransferases (GlcNAcTs) are glycosylating enzymes that play a key role in adding N-acetylglucosamine (GlcNAc) to substrates to facilitate their proper trafficking, intracellular targeting, and cellular function. Given previous results indicating abnormal glycosylation in schizophrenia, we hypothesized that these GlcNAcTs may be abnormally expressed in this illness. We measured protein expression of nine distinct GlcNAcTs by Western blot analysis in postmortem samples of dorsolateral prefrontal cortex (DLPFC) from twelve pairs of elderly patients with schizophrenia and comparison subjects. We found decreased protein expression of UDP-GlcNAc:BetaGal Beta-1,3 GlcNAcT 8 (B3GNT8) and mannosyl (alpha-1,3-)-glycoprotein beta-1,4 GlcNAcT (MGAT4A) expression in schizophrenia. These data provide further evidence that glycosylation is dysregulated in schizophrenia, and suggest a potential mechanism associated with alterations in protein function, trafficking, and intracellular targeting in this illness.
Sigma-Aldrich
Anti-MGAT4C antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)