Merck
  • Effect of Lycoris chejuensis and Its Active Components on Experimental Models of Alzheimer's Disease.
Effect of Lycoris chejuensis and Its Active Components on Experimental Models of Alzheimer's Disease.
Journal of agricultural and food chemistry (2015-07-29)
Joonki Kim, Yurim Park, Yoon Sun Chun, Jin Wook Cha, Hak Cheol Kwon, Myung Sook Oh, Sungkwon Chung, Hyun Ok Yang
ABSTRACT
We found that an extract of Lycoris chejuensis and its three isolated active components, narciclasine, 7-deoxynarciclasine, and 7-deoxy-trans-dihydronarciclasine, each significantly reduced the formation of amyloid-β peptides in HeLa cells transfected with an amyloid precursor protein carrying the Swedish mutation up to 45 ± 3.6%. The extract down-regulated amyloid precursor protein, especially the mature form by up to 88%, and reduced the ability of secretases to generate toxic amyloid-β. Double-transgenic mice treated with the extract for 4 months also showed significantly reduced levels of amyloid-β and plaques while exhibiting improved memory functions in the Morris water maze and novel object recognition tests. In conclusion, the extract and isolated active components of L. chejuensis decreased the production of amyloid-β by attenuating amyloid precursor protein levels. Furthermore, the extract improved the disrupted memory functions in animals while inhibiting amyloid plaque formation. Thus, this extract, as well as its active components, could prove beneficial in the treatment of Alzheimer's disease.
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%