Merck
  • Shape-controlled gold nanoparticles supported on MoS₂ nanosheets: synergistic effect of thionine and MoS₂ and their application for electrochemical label-free immunosensing.
Shape-controlled gold nanoparticles supported on MoS₂ nanosheets: synergistic effect of thionine and MoS₂ and their application for electrochemical label-free immunosensing.
Nanoscale (2015-11-03)
Shao Su, Min Zou, He Zhao, Chengfeng Yuan, Yanan Xu, Chi Zhang, Lihua Wang, Chunhai Fan, Lianhui Wang
ABSTRACT
Herein, a facile approach for shape-controlled gold nanoparticle (AuNP) decorated thionine-MoS2 nanocomposites (AuNP-Thi-MoS2) had been reported by using the synergistic effect of MoS2 and Thi. Thi was not only used as an electrochemical indicator, but also as a reducing agent to tune the resulting morphologies of AuNPs. With the ratio of MoS2/Thi decreasing, the shapes of AuNPs changed from spherical, triangle, clover-like to flower-like nanostructures. TEM, XRD and XPS were employed to characterize the formation and chemical composition of AuNP-Thi-MoS2 nanocomposites. On the basis of synthesis, a MoS2-based label-free electrochemical immunosensor had been designed to detect carcino-embryonic antigen (CEA). The proposed immunosensor could detect as low as 0.52 pg mL(-1) CEA with excellent selectivity. Moreover, the expected immunosensor showed high stability and excellent reproducibility, which could detect CEA in human serum with satisfactory results. Therefore, the AuNP-Thi-MoS2 nanocomposites may be considered as a candidate sensing platform for fabrication of simple, label-free and ultrasensitive electrochemical sensors.
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Thionin acetate salt, certified by the Biological Stain Commission, Dye content ≥85 %
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(III), 99.98% trace metals basis
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ACS reagent, 98.5-102.0%
Sigma-Aldrich
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O